Iron(II) phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation
Leverandør: TCI
Synonymer:
Phthalocyanine Iron(II), Iron phthalocyanine
I0783-200MG
I0783-1G
TCIAI0783-200MGEA
715
NOK
TCIAI0783-200MG
TCIAI0783-1G
Iron(II) phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation
Iron(II) phthalocyanine
Formel:
C₃₂H₁₆FeN₈ MW: 568,38 g/mol |
MDL nummer:
MFCD00015953 CAS-nummer: 132-16-1 EINECS: 205-047-2 |
Spesifikasjon Testresultater
Appearance | Dark purple powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...