Isopropylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

I0787-5G I0787-25G
TCIAI0787-5GEA 660 NOK
TCIAI0787-5G TCIAI0787-25G
Isopropylboronic acid (contains varying amounts of Anhydride)
Isopropylboronic acid
Formel: C₃H₉BO₂
MW: 87,91 g/mol
Smeltepunkt: 115°C
MDL nummer: MFCD01319021
CAS-nummer: 80041-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51b5a11610c4aa947ec6c7307195fd6b4e808273&vid=51b5a11610c4aa947ec6c7307195fd6b4e808273&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 96.0 to 126.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR