Isoamyl 4-methoxycinnamate ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
4-Methoxycinnamic Acid Isoamyl Ester, 4-Methoxycinnamic Acid Isopentyl Ester, Amiloxate, Isopentyl 4-Methoxycinnamate, Isopentyl 3-(4-methoxyphenyl)acrylate
I0815-5G
I0815-25G
TCIAI0815-5GEA
460
NOK
TCIAI0815-5G
TCIAI0815-25G
Isoamyl 4-methoxycinnamate ≥95.0% (by GC)
Isoamyl 4-methoxycinnamate
Formel:
C₁₅H₂₀O₃ MW: 248,32 g/mol Kokepunkt: 158°C |
MDL nummer:
MFCD00583856 CAS-nummer: 71617-10-2 EINECS: 275-702-5 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...