Icariin ≥96.0% (by HPLC)
Leverandør: TCI
Synonymer:
5-Hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
I0862-200MG
I0862-1G
TCIAI0862-200MGEA
525
NOK
TCIAI0862-200MG
TCIAI0862-1G
Icariin ≥96.0% (by HPLC)
Icariin
Formel:
C₃₃H₄₀O₁₅ MW: 676,67 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00210516 CAS-nummer: 489-32-7 Merck Indeks: 14,03617 |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Melting point | 235.0 to 239.0 deg-C |
Specific rotation [a]20/D | -82.0 to -88.0 deg (C=1,Pyridine) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...