3-Iodobenzyl alcohol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (3-Iodophenyl)methanol, 3-Iodobenzylalcohol

I0866-5G I0866-25G
TCIAI0866-5GEA 500 NOK
TCIAI0866-5G TCIAI0866-25G
3-Iodobenzyl alcohol ≥98.0% (by GC)
3-Iodobenzyl alcohol
Formel: C₇H₇IO
MW: 234,04 g/mol
Kokepunkt: 105°C
MDL nummer: MFCD00004635
CAS-nummer: 57455-06-8
EINECS: 260-744-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbf8c6ee0c0853dba6b75a91e885c8ef4e4a7b19&vid=cbf8c6ee0c0853dba6b75a91e885c8ef4e4a7b19&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR