Isopropyl-3-aminobut-2-enoate ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: Isopropyl 3-Aminocrotonate, Isopropyl 3-aminobut-2-enoate, 3-Aminocrotonic acid Isopropyl ester, Isopropyl 3-Amino-2-butenoate, 3-Amino-2-butenoic acid Isopropyl ester
I0884-1G I0884-5G
TCIAI0884-1GEA 310 NOK
TCIAI0884-1G TCIAI0884-5G
Isopropyl-3-aminobut-2-enoate ≥98.0% (by GC, titration analysis)
Isopropyl-3-aminobut-2-enoate
Formel: C₇H₁₃NO₂
MW: 143,1836 g/mol
Kokepunkt: 80°C
MDL nummer: MFCD00134272
CAS-nummer: 14205-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a62cacb09e0d1278974a72e6f56ea23dbd774921&vid=a62cacb09e0d1278974a72e6f56ea23dbd774921&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %