4-Iodo-1-tritylimidazole ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
4-Iodo-1-trityl-1H-imidazole
I0890-1G
I0890-5G
TCIAI0890-1GEA
485
NOK
TCIAI0890-1G
TCIAI0890-5G
4-Iodo-1-tritylimidazole ≥98.0% (by HPLC, titration analysis)
4-Iodo-1-tritylimidazole
Formel:
C₂₂H₁₇IN₂ MW: 436,2900 g/mol |
MDL nummer:
MFCD02179542 CAS-nummer: 96797-15-8 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 221.0 to 225.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...