5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide mixture of isomers ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: Iohexol, N1,N3-Bis(2,3-dihydroxypropyl)-5-(N-(2,3-dihydroxypropyl)acetamido)-2,4,6-triiodoisophthalamide, 5-[Acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide, N,N'-Bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodoisophthalamide

I0903-5G I0903-25G
TCIAI0903-5GEA 365 NOK
TCIAI0903-5G TCIAI0903-25G
5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide mixture of isomers ≥98.0% (by HPLC, total nitrogen)
5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide
Formel: C₁₉H₂₆I₃N₃O₉
MW: 821,142 g/mol
Smeltepunkt: 180°C
MDL nummer: MFCD00077732
CAS-nummer: 66108-95-0
EINECS: 266-164-2
Merck Indeks: 14,05052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02486f8ad5dd0c83549acdd9fc03c670db56200d&vid=02486f8ad5dd0c83549acdd9fc03c670db56200d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of isomers)
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR