1-Isochromanone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3,4-Dihydro-1H-2-benzopyran-1-one, Isochroman-1-one
I0953-1G
I0953-5G
TCIAI0953-1GEA
540
NOK
TCIAI0953-1G
TCIAI0953-5G
1-Isochromanone ≥98.0% (by GC)
1-Isochromanone
Formel:
C₉H₈O₂ MW: 148,16 g/mol Kokepunkt: 160°C |
MDL nummer:
MFCD00799232 CAS-nummer: 4702-34-5 EINECS: 225-179-4 |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...