Isoindoline hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

I0958-1G I0958-5G
TCIAI0958-1GEA 230 NOK
TCIAI0958-1G TCIAI0958-5G
Isoindoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Isoindoline hydrochloride
Formel: C₈H₁₀ClN
MDL nummer: MFCD03502377
CAS-nummer: 32372-82-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0ec0adb047ff9f29af7f382aa4c35fc8c87f288&vid=e0ec0adb047ff9f29af7f382aa4c35fc8c87f288&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 259.0 to 263.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR