5'-Iodo-m-terphenyl ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 1-Iodo-3,5-diphenylbenzene, 5'-Iodo-1,1':3',1''-terphenyl

I0969-200MG I0969-1G
TCIAI0969-200MGEA 850 NOK
TCIAI0969-200MG TCIAI0969-1G
5'-Iodo-m-terphenyl ≥98.0% (by GC)
5'-Iodo-m-terphenyl
Formel: C₁₈H₁₃I
MW: 356,21 g/mol
MDL nummer: MFCD26131291
CAS-nummer: 87666-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c909eb3658a429f0cb5ead433684c175fec1eac&vid=1c909eb3658a429f0cb5ead433684c175fec1eac&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR