4-Iodo-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Leverandør: TCI
Danger

I1018-1G I1018-5G
TCIAI1018-1GEA 435 NOK
TCIAI1018-1G TCIAI1018-5G
4-Iodo-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Iodo-2-(trifluoromethyl)benzonitrile
Formel: C₈H₃F₃IN
MW: 297,02 g/mol
MDL nummer: MFCD08458091
CAS-nummer: 101066-87-9
FN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be799a61eea37261bd5981ec42acd962b829583b&vid=be799a61eea37261bd5981ec42acd962b829583b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR