Inosine Pranobex ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
4-Acetamidobenzoic acid,9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one,1-(Dimethylamino)propan-2-ol (3:1:3)
I1037-100MG
I1037-1G
TCIAI1037-100MGEA
260
NOK
TCIAI1037-100MG
TCIAI1037-1G
Inosine Pranobex ≥98.0% (by HPLC, titration analysis)
Inosine Pranobex
Formel:
C₁₀H₁₂N₄O₅·₃C₉H₉NO₃·3C₅H₁₃NO MW: 1115,25 g/mol |
MDL nummer:
MFCD05662374 CAS-nummer: 36703-88-5 EINECS: 253-162-1 Merck Indeks: 14,04976 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -9.0 to -13.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...