1H-Indole-3-carbaldehyde oxime ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1H-Indole-3-carbaldehyde Oxime, 1H-Indole-3-carboxaldehyde Oxime

I1060-1G I1060-5G
TCIAI1060-1GEA 1125 NOK
TCIAI1060-1G TCIAI1060-5G
1H-Indole-3-carbaldehyde oxime ≥98.0% (by GC)
1H-Indole-3-carbaldehyde oxime
Formel: C₉H₈N₂O
MW: 160,18 g/mol
MDL nummer: MFCD00022737
CAS-nummer: 2592-05-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR