Isosteviol ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: (4alpha,8beta,13beta)-13-Methyl-16-oxo-17-norkauran-18-oic acid, (4R,4aS,6aR,9S,11aR,11bS)-4,9,11b-Trimethyl-8-oxotetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylic acid

I1069-100MG I1069-500MG
TCIAI1069-100MGEA 1175 NOK
TCIAI1069-100MG TCIAI1069-500MG
Isosteviol ≥98.0% (by GC, titration analysis)
Isosteviol
Formel: C₂₀H₃₀O₃
MW: 318,46 g/mol
MDL nummer: MFCD28411514
CAS-nummer: 27975-19-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 228.0 to 232.0 deg-C
Specific rotation [a]20/D -78.0 to -82.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR