4-Isopropylbenzylamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: (4-Isopropylphenyl)methanamine

I1074-1ML I1074-5ML
TCIAI1074-1MLEA 700 NOK
TCIAI1074-1ML TCIAI1074-5ML
4-Isopropylbenzylamine ≥98.0% (by GC, titration analysis)
4-Isopropylbenzylamine
Formel: C₁₀H₁₅N
MW: 149,24 g/mol
Kokepunkt: 227°C
MDL nummer: MFCD00025580
CAS-nummer: 4395-73-7
FN: 2735
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR