Isopropyl linoleate >60.0% (by GC)
Leverandør: TCI
Synonymer:
Linoleic Acid Isopropyl Ester
L0139-25ML
L0139-500ML
TCIAL0139-25MLEA
220
NOK
TCIAL0139-25ML
TCIAL0139-500ML
Isopropyl linoleate >60.0% (by GC)
Isopropyl linoleate
Formel:
C₂₁H₃₈O₂ MW: 322,53 g/mol Kokepunkt: 179°C Flash punkt: 191°C |
MDL nummer:
MFCD00053809 CAS-nummer: 22882-95-7 EINECS: 245-289-6 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 60.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...