L-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen basis)
Leverandør: TCI
Synonymer:
H-Lys-OMe·2HCl, H-Lys-OMe.2HCl
L0202-25G
L0202-5G
TCIAL0202-25GEA
1225
NOK
TCIAL0202-25G
TCIAL0202-5G
L-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen basis)
L-Lysine methyl ester dihydrochloride
Formel:
C₇H₁₆N₂O₂ MW: 233,13 g/mol |
MDL nummer:
MFCD00039067 CAS-nummer: 26348-70-9 EINECS: 247-625-7 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | +13.0 to +19.0 deg (C=5, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...