Levosimendan ≥98.0% (by HPLC)

Leverandør: TCI
Danger

Synonymer: OR-1259, 2-[2-[4-[(4R)-1,4,5,6-Tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazinylidene]propanedinitrile, (R)-N-(4-(4-Methyl-6-oxo-1,4,5,6-tetrahydropyridazin-3-yl)phenyl)carbonohydrazonoyl dicyanide, (R)-Simendan

L0320-50MG L0320-250MG
TCIAL0320-50MGEA 835 NOK
TCIAL0320-50MG TCIAL0320-250MG
Levosimendan ≥98.0% (by HPLC)
Levosimendan
Formel: C₁₄H₁₂N₆O
MW: 280,29 g/mol
MDL nummer: MFCD00867135
CAS-nummer: 141505-33-1
FN: 3439
ADR: 6.1,III
Merck Indeks: 14,05471

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -600 to -650 deg (C=0.5, THF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR