Dibutyl-DL-malate ≥95.0% (by GC)

Leverandør: TCI

Synonymer: DL-Apple Acid Dibutyl Ester, DL-Malic Acid Dibutyl Ester

M0023-25G M0023-500G
TCIAM0023-25GEA 905 NOK
TCIAM0023-25G TCIAM0023-500G
Dibutyl-DL-malate ≥95.0% (by GC)
Dibutyl-DL-malate
Formel: C₁₂H₂₂O₅
MW: 246.30 g/mol
MDL nummer: MFCD00059251
CAS-nummer: 6280-99-5
EINECS: 228-485-6

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR