Diethyl-DL-malate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: DL-Malic Acid Diethyl Ester, Diethyl 2-hydroxysuccinate, DL-Apple Acid Diethyl Ester

M0024-25G M0024-500G
TCIAM0024-25GEA 350 NOK
TCIAM0024-25G TCIAM0024-500G
Diethyl-DL-malate ≥98.0% (by GC)
Diethyl-DL-malate
Formel: C₈H₁₄O₅
MW: 190,20 g/mol
Kokepunkt: 255°C
Flash punkt: 85°C
MDL nummer: MFCD00048556
CAS-nummer: 7554-12-3
EINECS: 231-444-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR