4,4'-Methylenebis(2,6-di-tert-butylphenol) ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Bis(3,5-di-tert-butyl-4-hydroxyphenyl)methane

M0214-25G M0214-500G
TCIAM0214-25GEA 310 NOK
TCIAM0214-25G TCIAM0214-500G
4,4'-Methylenebis(2,6-di-tert-butylphenol) ≥98.0% (by GC)
4,4'-Methylenebis(2,6-di-tert-butylphenol)
Formel: C₂₉H₄₄O₂
MW: 424,67 g/mol
MDL nummer: MFCD00008822
CAS-nummer: 118-82-1
EINECS: 204-279-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 154.0 to 158.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR