Methylenedisalicylic acid (mixture of isomers) ≥90.0% (by titrimetric analysis)

Leverandør: TCI
Warning

M0221-25G M0221-500G
TCIAM0221-25GEA 220 NOK
TCIAM0221-25G TCIAM0221-500G
Methylenedisalicylic acid (mixture of isomers) ≥90.0% (by titrimetric analysis)
Methylenedisalicylic acid (mixture of isomers)
Formel: C₁₅H₁₂O₆
MDL nummer: MFCD00016506
CAS-nummer: 27496-82-8
EINECS: 248-497-5
Merck Indeks: 14,06065

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light red to Green powder to crystal
Purity (Neutralization titration) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR