3-Carbamyl-1-methylpyridinium chloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-Carbamoyl-1-methylpyridinium Chloride, TRIA-662, 3-Carbamoyl-1-methylpyridin-1-ium chloride

M0375-25G M0375-5G
TCIAM0375-25GEA 3300 NOK
TCIAM0375-25G TCIAM0375-5G
3-Carbamyl-1-methylpyridinium chloride ≥98.0% (by HPLC, titration analysis)
3-Carbamyl-1-methylpyridinium chloride
Formel: C₇H₉ClN₂O
MW: 172,61 g/mol
MDL nummer: MFCD00060042
CAS-nummer: 1005-24-9
EINECS: 463-670-7
Merck Indeks: 14,09693

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR