Isobutyl myristate ≥97.0% (by GC)
Leverandør: TCI
Synonymer:
Myristicacid isobutylester, Myristic Acid Isobutyl Ester
M0555-25ML
M0555-500ML
TCIAM0555-25MLEA
1275
NOK
TCIAM0555-25ML
TCIAM0555-500ML
Isobutyl myristate ≥97.0% (by GC)
Isobutyl myristate
Formel:
C₁₈H₃₆O₂ MW: 284,48 g/mol Flash punkt: 152°C |
MDL nummer:
MFCD00059286 CAS-nummer: 25263-97-2 EINECS: 246-765-6 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...