Monomethyl maleate ≥90.0% (by GC)
Leverandør: TCI
Warning
Synonymer: Methyl Hydrogen Maleate, (Z)-4-Methoxy-4-oxobut-2-enoic acid, Maleic Acid Monomethyl Ester
M0624-25ML M0624-500ML M0624-100ML
TCIAM0624-25MLEA 540 NOK
TCIAM0624-25ML TCIAM0624-500ML TCIAM0624-100ML
Monomethyl maleate ≥90.0% (by GC)
Monomethyl maleate
Formel: C₅H₆O₄
MW: 130,10 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00045905
CAS-nummer: 3052-50-4
EINECS: 221-271-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d25baa348ca09251af4504f82e346e3e5b79d263&vid=d25baa348ca09251af4504f82e346e3e5b79d263&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 90.0 %