3-(Methylthio)propionyl chloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Danger
Synonymer: 3-(Methylmercapto)propionyl Chloride
M0806-25ML M0806-500ML
TCIAM0806-25MLEA 985 NOK
TCIAM0806-25ML TCIAM0806-500ML
3-(Methylthio)propionyl chloride ≥98.0% (by titrimetric analysis)
3-(Methylthio)propionyl chloride
Formel: C₄H₇ClOS
MW: 138,61 g/mol
Kokepunkt: 96°C
MDL nummer: MFCD00059482
CAS-nummer: 7031-23-4
FN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=796cfb7da7e3e2fb6cb39c8f6998ebaac6ee7e5b&vid=796cfb7da7e3e2fb6cb39c8f6998ebaac6ee7e5b&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 98.0 %