Methyl-1-methylpyrrole-2-acetate ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Methyl 2-(1-methyl-1H-pyrrol-2-yl)acetate, 1-Methylpyrrole-2-acetic Acid Methyl Ester
M0846-25ML
M0846-5ML
TCIAM0846-25MLEA
1650
NOK
TCIAM0846-25ML
TCIAM0846-5ML
Methyl-1-methylpyrrole-2-acetate ≥97.0% (by GC)
Methyl-1-methylpyrrole-2-acetate
Formel:
C₈H₁₁NO₂ MW: 153,18 g/mol Kokepunkt: 85°C Flash punkt: 106°C |
MDL nummer:
MFCD00009752 CAS-nummer: 51856-79-2 EINECS: 257-478-0 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...