2-Hydroxy-N-methylbenzylamine hydrochloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
N-Methyl-2-hydroxybenzylamine Hydrochloride, 2-(Methylaminomethyl)phenol Hydrochloride, alpha-Methylamino-o-cresol Hydrochloride
M1005-5G
M1005-1G
TCIAM1005-5GEA
2550
NOK
TCIAM1005-5G
TCIAM1005-1G
2-Hydroxy-N-methylbenzylamine hydrochloride ≥98.0% (by titrimetric analysis)
2-Hydroxy-N-methylbenzylamine hydrochloride
Formel:
C₈H₁₂NO MW: 173,64 g/mol |
MDL nummer:
MFCD00142934 CAS-nummer: 60399-02-2 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...