2-Methyl-1-phenylpropene ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (2-Methylprop-1-en-1-yl)benzene, (2-Methylpropenyl)benzene

M1015-1G M1015-5G
TCIAM1015-1GEA 1225 NOK
TCIAM1015-1G TCIAM1015-5G
2-Methyl-1-phenylpropene ≥98.0% (by GC)
2-Methyl-1-phenylpropene
Formel: C₁₀H₁₂
MW: 132,21 g/mol
Kokepunkt: 189°C
Flash punkt: 59°C
MDL nummer: MFCD00008899
CAS-nummer: 768-49-0
EINECS: 212-194-6
FN: 3295
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR