(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid ≥99.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
Naproxen, (S)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid
M1021-25G
M1021-500G
TCIAM1021-25GEA
810
NOK
TCIAM1021-25G
TCIAM1021-500G
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid ≥99.0% (by GC, titration analysis)
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Formel:
C₁₄H₁₄O₃ MW: 230,26 g/mol |
MDL nummer:
MFCD00010500 CAS-nummer: 22204-53-1 EINECS: 244-838-7 FN: 2811 ADR: 6.1,III Merck Indeks: 14,06417 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 155.0 to 158.0 deg-C |
Specific rotation [a]20/D | +65.0 to +70.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...