(3S)-(-)-3-(Methylamino)pyrrolidine ≥97.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
M1108-1G
M1108-5G
TCIAM1108-1GEA
2375
NOK
TCIAM1108-1G
TCIAM1108-5G
(3S)-(-)-3-(Methylamino)pyrrolidine ≥97.0% (by GC, titration analysis)
(3S)-(-)-3-(Methylamino)pyrrolidine
Formel:
C₅H₁₂N₂ MW: 100,17 g/mol Kokepunkt: 164°C |
MDL nummer:
MFCD00191462 CAS-nummer: 139015-32-0 FN: 2734 ADR: 8,II |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Specific rotation [a]20/D | -17.0 to -22.0 deg (C=10, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...