3,7-Dimethyloctane ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
D1214-1ML
D1214-5ML
TCIAD1214-1MLEA
1650
NOK
TCIAD1214-1ML
TCIAD1214-5ML
3,7-Dimethyloctane ≥98.0% (by GC)
3,7-Dimethyloctane
Formel:
C₁₀H₂₂ Kokepunkt: 160°C Smeltepunkt: –100 °C Tetthet: 0,7139 g/cm³ (20 °C) Flash punkt: 38°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00048739 CAS-nummer: 15869-93-9 FN: 3295 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...