2-Methoxyethyl methanesulphonate ≥97.0% (by GC)

Leverandør: TCI

Synonymer: Methanesulphonic Acid 2-Methoxyethyl Ester

M1446-25G M1446-5G
TCIAM1446-25GEA 2325 NOK
TCIAM1446-25G TCIAM1446-5G
2-Methoxyethyl methanesulphonate ≥97.0% (by GC)
2-Methoxyethyl methanesulphonate
Formel: C₄H₁₀O₄S
MW: 154,18 g/mol
Kokepunkt: 126°C
MDL nummer: MFCD02656401
CAS-nummer: 16427-44-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR