ɑ-Methylene-γ-valerolactone ≥97.0% (by GC) stabilised

Leverandør: TCI

Synonymer: 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone

Stabilisering: stabilisert med hydroquinon

M1453-25G M1453-5G
TCIAM1453-25GEA 4625 NOK
TCIAM1453-25G TCIAM1453-5G
ɑ-Methylene-γ-valerolactone ≥97.0% (by GC) stabilised
alpha-Methylene-gamma-valerolactone
Formel: C₆H₈O₂
MW: 112,13 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06797118
CAS-nummer: 62873-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4e926c75aac8a915077f917ea79c32e7f534d25&vid=a4e926c75aac8a915077f917ea79c32e7f534d25&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR