N-Methyl-p-toluenesulphonamide ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: N,4-Dimethylbenzenesulphonamide

M1717-25G M1717-250G
TCIAM1717-25GEA 405 NOK
TCIAM1717-25G TCIAM1717-250G
N-Methyl-p-toluenesulphonamide ≥98.0% (by HPLC, total nitrogen)
N-Methyl-p-toluenesulphonamide
Formel: C₈H₁₁NO₂S
MW: 185,24 g/mol
MDL nummer: MFCD00008285
CAS-nummer: 640-61-9
EINECS: 211-366-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 77.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR