Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate ≥98.0% (by GC)
Leverandør: TCI
|
|
Synonymer:
Methyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid methyll ester, Methyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
M1810-5G
M1810-25G
TCIAM1810-5GEA
260
NOK
TCIAM1810-5G
TCIAM1810-25G
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate ≥98.0% (by GC)
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate
Formel:
C₁₈H₂₈O₃ MW: 292,42 g/mol Kokepunkt: 130°C |
MDL nummer:
MFCD00210461 CAS-nummer: 6386-38-5 EINECS: 228-985-4 FN: 3077 ADR: 9,III |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...