2-Methyl-3-(methyldithio)furan ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-Methyl-3-(methyldisulphanyl)furan, Methyl 2-Methyl-3-furyl Disulphide

M1837-1G M1837-5G
TCIAM1837-1GEA 380 NOK
TCIAM1837-1G TCIAM1837-5G
2-Methyl-3-(methyldithio)furan ≥98.0% (by GC)
2-Methyl-3-(methyldithio)furan
Formel: C₆H₈OS₂
MW: 160,25 g/mol
Kokepunkt: 37°C
Flash punkt: 84°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00038643
CAS-nummer: 65505-17-1
EINECS: 265-797-1
FN: 2810
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR