2-Methyl-3-(methylthio)furan ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
M1838-25G
M1838-5G
TCIAM1838-25GEA
2100
NOK
TCIAM1838-25G
TCIAM1838-5G
2-Methyl-3-(methylthio)furan ≥98.0% (by GC)
2-Methyl-3-(methylthio)furan
Formel:
C₆H₈OS MW: 128,19 g/mol Flash punkt: 59°C |
MDL nummer:
MFCD03701635 CAS-nummer: 63012-97-5 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...