Methyl trans-2-octenoate ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
trans-2-Octenoic Acid Methyl Ester, Methyl trans-2-Octenoate, (E)-2-Methyl oct-2-enoate
M2014-25ML
M2014-5ML
TCIAM2014-25MLEA
4150
NOK
TCIAM2014-25ML
TCIAM2014-5ML
Methyl trans-2-octenoate ≥95.0% (by GC)
Methyl trans-2-octenoate
Formel:
C₉H₁₆O₂ MW: 156,23 g/mol Kokepunkt: 91°C Flash punkt: 83°C |
MDL nummer:
MFCD00036620 CAS-nummer: 7367-81-9 EINECS: 230-912-6 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...