5-Methylpyrazine-2-carboxylic acid-4-oxide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Acipimox, 5-Carboxy-2-methylpyrazine 1-oxide, 2-Carboxy-5-methylpyrazine 4-Oxide, 5-Carboxy-2-methylpyrazine 1-Oxide

M2053-1G M2053-5G
TCIAM2053-1GEA 745 NOK
TCIAM2053-1G TCIAM2053-5G
5-Methylpyrazine-2-carboxylic acid-4-oxide ≥98.0% (by GC)
5-Methylpyrazine-2-carboxylic acid-4-oxide
Formel: C₆H₆N₂O₃
MW: 154,13 g/mol
Smeltepunkt: 180°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00865757
CAS-nummer: 51037-30-0
EINECS: 256-928-3
Merck Indeks: 14,00111

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR