4-Methoxy-4'-methyldiphenylamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-Methoxy-N-(p-tolyl)aniline, N-(4-Methoxyphenyl)-p-tolylamine

M2070-25G M2070-5G
TCIAM2070-25GEA 3150 NOK
TCIAM2070-25G TCIAM2070-5G
4-Methoxy-4'-methyldiphenylamine ≥98.0% (by GC, titration analysis)
4-Methoxy-4'-methyldiphenylamine
Formel: C₁₄H₁₅NO
MW: 213,28 g/mol
MDL nummer: MFCD00092922
CAS-nummer: 39253-43-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR