1-(Mercaptomethyl)cyclopropaneacetic acid ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: [1-(Thiomethyl)cycloprop-1-yl]acetic acid, 2-(1-(Mercaptomethyl)cyclopropyl)acetic acid

M2074-1G M2074-5G
TCIAM2074-1GEA 690 NOK
TCIAM2074-1G TCIAM2074-5G
1-(Mercaptomethyl)cyclopropaneacetic acid ≥97.0% (by GC, titration analysis)
1-(Mercaptomethyl)cyclopropaneacetic acid
Formel: C₆H₁₀O₂S
MW: 146,20 g/mol
MDL nummer: MFCD03411600
CAS-nummer: 162515-68-6
EINECS: 420-240-3
FN: 3261
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR