5-(Trifluoromethyl)-m-anisidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-Amino-5-(trifluoromethyl)anisole, 3-Amino-5-methoxybenzotrifluoride, 3-Methoxy-5-(trifluoromethyl)aniline, 5-(Trifluoromethyl)-3-anisidine, 5-(Trifluoromethyl)-3-methoxyaniline

M2262-1G M2262-5G
TCIAM2262-1GEA 485 NOK
TCIAM2262-1G TCIAM2262-5G
5-(Trifluoromethyl)-m-anisidine ≥98.0% (by GC, titration analysis)
5-(Trifluoromethyl)-m-anisidine
Formel: C₈H₈F₃NO
MW: 191,15 g/mol
MDL nummer: MFCD00000393
CAS-nummer: 349-55-3
EINECS: 206-487-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cd89fc11fa56fa631dab3f367f8138ed4de7b65&vid=6cd89fc11fa56fa631dab3f367f8138ed4de7b65&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR