2-Mercapto-5-(trifluoromethyl)pyridine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 5-(Trifluoromethyl)-2(1H)-pyridinethione, 5-(Trifluoromethyl)-2-pyridinethiol, 5-(Trifluoromethyl)pyridine-2(1H)-thione

M2299-5G M2299-25G
TCIAM2299-5GEA 1650 NOK
TCIAM2299-5G TCIAM2299-25G
2-Mercapto-5-(trifluoromethyl)pyridine ≥98.0% (by HPLC, titration analysis)
2-Mercapto-5-(trifluoromethyl)pyridine
Formel: C₆H₄F₃NS
MW: 179,16 g/mol
Smeltepunkt: 150°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00128893
CAS-nummer: 76041-72-0

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR