2'-O-Methylcytidine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 2′-O-methylcytidine

M2317-200MG M2317-1G
TCIAM2317-200MGEA 485 NOK
TCIAM2317-200MG TCIAM2317-1G
2'-O-Methylcytidine ≥98.0% (by HPLC, titration analysis)
2'-O-Methylcytidine
Formel: C₁₀H₁₅N₃O₅
MW: 257,25 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00056067
CAS-nummer: 2140-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5e071eb47cc1db820f3e59d1256945e40d155c7&vid=a5e071eb47cc1db820f3e59d1256945e40d155c7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +70.0 to +74.0 deg (C=1、H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR