3-Methyl-6-nitro-1H-indazole ≥98.0% (by GC)

Leverandør: TCI
Warning

M2358-5G M2358-25G
TCIAM2358-5GEA 245 NOK
TCIAM2358-5G TCIAM2358-25G
3-Methyl-6-nitro-1H-indazole ≥98.0% (by GC)
3-Methyl-6-nitro-1H-indazole
Formel: C₈H₇N₃O₂
MW: 177,1600 g/mol
MDL nummer: MFCD07082704
CAS-nummer: 6494-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73eb4a65c8c98efcab6a2d051d0f8445f0307bbd&vid=73eb4a65c8c98efcab6a2d051d0f8445f0307bbd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 184.0 to 188.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR