Methoxycyclohexane ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Cyclohexyl methyl ether

M2374-5G M2374-25G
TCIAM2374-5GEA 1550 NOK
TCIAM2374-5G TCIAM2374-25G
Methoxycyclohexane ≥98.0% (by GC)
Methoxycyclohexane
Formel: C₇H₁₄O
MW: 114,19 g/mol
Kokepunkt: 135°C
Smeltepunkt: –74°C
MDL nummer: MFCD00154087
CAS-nummer: 931-56-6
FN: 3271
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52c069f3af6ee0398ba0e2f5d9726b9aed230687&vid=52c069f3af6ee0398ba0e2f5d9726b9aed230687&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR