Mycophenolate mofetil ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 2-Morpholinoethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoate, 2-(4-Morpholinyl)ethyl (E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoate, (E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic Acid 2-(4-Morpholinyl)ethyl Ester

M2387-100MG M2387-1G
TCIAM2387-100MGEA 245 NOK
TCIAM2387-100MG TCIAM2387-1G
Mycophenolate mofetil ≥98.0% (by HPLC, titration analysis)
Mycophenolate mofetil
Formel: C₂₃H₃₁NO₇
MW: 433,50 g/mol
MDL nummer: MFCD00867568
CAS-nummer: 128794-94-5
Merck Indeks: 14,06327
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=531c09d3a48874f9c316cd024f4e6ee6af1bd9c1&vid=531c09d3a48874f9c316cd024f4e6ee6af1bd9c1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 94.0 to 98.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR