4-(Methylsulphonyl)benzyl bromide ≥97.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
α-Bromo-4-(methylsulphonyl)toluene, 1-(Bromomethyl)-4-(methylsulphonyl)benzene
M2474-1G
M2474-5G
TCIAM2474-1GEA
905
NOK
TCIAM2474-1G
TCIAM2474-5G
4-(Methylsulphonyl)benzyl bromide ≥97.0% (by GC, titration analysis)
4-(Methylsulphonyl)benzyl bromide
Formel:
C₈H₉BrO₂S MW: 249,13 g/mol |
MDL nummer:
MFCD00185849 CAS-nummer: 53606-06-7 FN: 3261 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...